siRNA Details
Gene Symbol: Syk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012758.1 | NP_036890.1 | 3 | 521 | ||
GenBank mRNA | L20838.1 | - | 1 | 157 | ||
U21683.1 | - | 3 | 521 | |||
U21684.1 | - | 3 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
