siRNA Details
Gene Symbol: Cdkn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130812.3 | NP_570825.1 | 2 | 638 | ||
GenBank mRNA | AF474978.1 | - | 2 | 552 | ||
AF474979.1 | - | 2 | 554 | |||
BC086995.1 | - | 2, 3 | 496 | 7134283 | ||
CK603883.1 | - | 2, 3 | 499 | |||
S79760.1 | - | 2 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
