siRNA Details
Gene Symbol: Ptpra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012763.2 | NP_036895.2 | 16 | 1976 | ||
XM_006235005.2 | XP_006235067.1 | 17 | 1763 | |||
XM_006235006.2 | XP_006235068.1 | 17 | 2039 | |||
XM_006235007.2 | XP_006235069.1 | 17 | 2557 | |||
XM_006235008.2 | XP_006235070.1 | 16 | 1736 | |||
GenBank mRNA | BC081828.1 | - | 16 | 1976 | 7107006 | |
L01702.1 | - | 15 | 1721 |

More Information
Other Species Targeted

Gene Ontology Categories:
