siRNA Details
Gene Symbol: Gdi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017088.2 | NP_058784.2 | 5 | 528 | ||
GenBank mRNA | AF130987.1 | - | 2 | 164 | ||
BC091156.1 | - | 5 | 532 | |||
BC099763.1 | - | 5 | 528 | 7930350 | ||
U07952.1 | - | 5 | 445 | |||
X74402.1 | - | 5 | 434 |

More Information
Other Species Targeted

Gene Ontology Categories:
