siRNA Details
Gene Symbol: Efnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017089.2 | NP_058785.2 | 3 | 1229 | ||
GenBank mRNA | BC061744.1 | - | 3 | 1229 | 5599058 | |
U07560.1 | - | 3 | 1255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
