siRNA Details
Gene Symbol: Klrb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173292.2 | NP_775414.2 | 3 | 207 | ||
GenBank mRNA | EF100682.1 | - | 3 | 207 | ||
EF100684.1 | - | 3 | 207 | |||
FJ158034.1 | - | 3 | 207 | |||
U56936.1 | - | 393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
