siRNA Details
Gene Symbol: Gnrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012767.2 | NP_036899.1 | 4 | 378 | ||
XM_006252096.2 | XP_006252158.1 | 3 | 379 | |||
GenBank mRNA | M12579.1 | - | 4 | 378 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
