siRNA Details
Gene Symbol: Ccnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC059113.1 | - | 5 | 702 | 6922392 | |
EV765751.1 | - | 1 | 107 | |||
FN800627.1 | - | 3 | 355 | |||
L11995.1 | - | 5 | 694 | |||
X60768.1 | - | 5 | 632 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
