siRNA Details
Gene Symbol: Tspy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022923.1 | NP_075212.1 | 3 | 866 | ||
XM_008773672.1 | XP_008771894.1 | 3 | 873 | |||
GenBank mRNA | AF074880.1 | - | 3 | 866 | ||
AJ001377.1 | - | 3 | 277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
