siRNA Details
Gene Symbol: Acvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022441.2 | NP_071886.1 | 9 | 1421 | ||
XM_006242303.2 | XP_006242365.1 | 10 | 1822 | |||
XM_006242304.2 | XP_006242366.1 | 10 | 2426 | |||
XM_006242305.2 | XP_006242367.1 | 9 | 2441 | |||
XM_006242307.2 | XP_006242369.1 | 9 | 2311 | |||
XM_008765673.1 | XP_008763895.1 | 10 | 2485 | |||
GenBank mRNA | BC083173.1 | - | 9 | 1421 | 7098096 | |
FQ223580.1 | - | 9 | 1463 | |||
L36088.1 | - | 10 | 1836 |
