siRNA Details
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012789.1 | NP_036921.1 | 11 | 1096 | ||
XM_008761879.1 | XP_008760101.1 | 12 | 1605 | |||
GenBank mRNA | J02997.1 | - | 11 | 1036 | ||
J04591.1 | - | 11 | 1096 |
