siRNA Details
Gene Symbol: Dyrk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012791.2 | NP_036923.1 | 5 | 555 | ||
XM_006248031.2 | XP_006248093.1 | 5 | 3500 | |||
XM_008768562.1 | XP_008766784.1 | 4 | 2763 | |||
XM_008768563.1 | XP_008766785.1 | 6 | 3636 | |||
XM_008768564.1 | XP_008766786.1 | 5 | 602 | |||
XM_008768565.1 | XP_008766787.1 | 5 | 677 | |||
GenBank mRNA | X79769.1 | - | 5 | 557 |
