siRNA Details
Gene Symbol: Itpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007235.2 | NP_001007236.2 | 43 | 5460 | ||
NM_001270596.1 | NP_001257525.1 | 42 | 5415 | |||
NM_001270597.1 | NP_001257526.1 | 40 | 5343 | |||
XM_006236987.2 | XP_006237049.1 | 43 | 5528 | |||
XM_008763167.1 | XP_008761389.1 | 43 | 5926 | |||
XM_008763168.1 | XP_008761390.1 | 43 | 5923 | |||
XM_008763169.1 | XP_008761391.1 | 43 | 5899 | |||
XM_008763170.1 | XP_008761392.1 | 43 | 5896 | |||
XM_008763171.1 | XP_008761393.1 | 42 | 5892 | |||
XM_008763172.1 | XP_008761394.1 | 42 | 5889 | |||
XM_008763173.1 | XP_008761395.1 | 42 | 5880 | |||
XM_008763174.1 | XP_008761396.1 | 41 | 5856 | |||
XM_008763175.1 | XP_008761397.1 | 42 | 5853 | |||
XM_008763176.1 | XP_008761398.1 | 40 | 5804 | |||
XM_008763177.1 | XP_008761399.1 | 42 | 5877 | |||
XM_008763178.1 | XP_008761400.1 | 39 | 5732 | |||
GenBank mRNA | GQ233032.1 | - | 41 | 5222 | ||
J05510.1 | - | 43 | 5554 | |||
M64698.1 | - | 2 | 157 | |||
M64699.1 | - | 5 | 277 | |||
U38653.1 | - | 3 | 232 | |||
U38665.1 | - | 6 | 349 |

More Information
Other Species Targeted

Gene Ontology Categories:
