siRNA Details
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012801.1 | NP_036933.1 | 6 | 784 | ||
XM_006248918.2 | XP_006248980.1 | 4 | 766 | |||
XM_006248919.2 | XP_006248981.1 | 4 | 766 | |||
XM_008768940.1 | XP_008767162.1 | 5 | 2365 | |||
GenBank mRNA | BC061731.1 | - | 4 | 771 | 5598667 | |
D10106.1 | - | 4 | 342 | |||
L06238.1 | - | 6 | 784 | |||
L06894.1 | - | 5 | 428 | |||
S57864.1 | - | 2 | 129 | |||
Z14120.1 | - | 4 | 278 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008808, BC003817, AY324648, AK017169, M29464.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
