siRNA Details
Gene Symbol: Sycp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012810.1 | NP_036942.1 | 13 | 1067 | ||
GenBank mRNA | X67805.1 | - | 13 | 1067 |

More Information
Other Species Targeted

Gene Ontology Categories:
