siRNA Details
Gene Symbol: Cox6a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012812.3 | NP_036944.2 | 3 | 311 | ||
XM_006230218.1 | XP_006230280.1 | 4 | 360 | |||
GenBank mRNA | FM058713.1 | - | 4 | 365 | ||
FM121690.1 | - | 3 | 217 | |||
FQ217951.1 | - | 3 | 261 | |||
FQ217987.1 | - | 3 | 218 | |||
FQ223608.1 | - | 3 | 267 | |||
FQ224015.1 | - | 3 | 269 | |||
FQ224196.1 | - | 3 | 268 | |||
FQ224467.1 | - | 3 | 257 | |||
FQ224572.1 | - | 3 | 276 | |||
X12554.1 | - | 3 | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
