siRNA Details
Gene Symbol: Nup153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100470.1 | NP_001093940.1 | 18 | 3309 | ||
XM_008771532.1 | XP_008769754.1 | 17 | 3092 | |||
GenBank mRNA | BC084700.1 | - | 1 | 98 | ||
CV117688.1 | - | 3 | 337 | |||
L06821.1 | - | 18 | 3309 |

More Information
Other Species Targeted

Gene Ontology Categories:
