siRNA Details
Gene Symbol: Cox6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012814.1 | NP_036946.1 | 3 | 305 | ||
GenBank mRNA | AA685532.1 | - | 1 | 12 | ||
CB716915.1 | - | 3 | 305 | |||
FQ211611.1 | - | 3 | 292 | |||
FQ213920.1 | - | 3 | 292 | |||
FQ221129.1 | - | 3 | 297 | |||
FQ221354.1 | - | 3 | 292 | |||
FQ221968.1 | - | 3 | 294 | |||
FQ224366.1 | - | 3 | 292 | |||
FQ225833.1 | - | 3 | 292 | |||
X12553.1 | - | 3 | 206 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007748, AK019318, AK002402, BC052816, U08440, and others.
Gene Ontology Categories:
