siRNA Details
Gene Symbol: Igfbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012817.1 | NP_036949.1 | 1 | 854 | ||
GenBank mRNA | BC087030.1 | - | 1056 | |||
M62781.1 | - | 1 | 854 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010518, L12447, AK142108, BC054812, BC057447, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
