siRNA Details
Gene Symbol: Acsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012820.1 | NP_036952.1 | 17 | 1735 | ||
XM_006253122.1 | XP_006253184.1 | 18 | 2395 | |||
XM_006253123.1 | XP_006253185.1 | 18 | 1936 | |||
XM_006253124.2 | XP_006253186.1 | 18 | 1855 | |||
XM_006253125.2 | XP_006253187.1 | 18 | 2015 | |||
XM_006253126.1 | XP_006253188.1 | 18 | 1861 | |||
XM_006253127.1 | XP_006253189.1 | 18 | 1826 | |||
XM_008771254.1 | XP_008769476.1 | 17 | 1761 | |||
GenBank mRNA | D90109.1 | - | 17 | 1735 | ||
FQ217275.1 | - | 17 | 1705 | |||
FQ219468.1 | - | 1 | 36 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
