siRNA Details
Gene Symbol: Apoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109996.1 | NP_001103466.1 | 3 | 161 | ||
NM_012824.2 | NP_036956.1 | 2 | 196 | |||
GenBank mRNA | AY327505.1 | - | 3 | 178 | ||
BC098670.1 | - | 2 | 135 | 7372566 | ||
FQ210440.1 | - | 3 | 161 | |||
FQ210757.1 | - | 3 | 150 | |||
FQ211211.1 | - | 3 | 162 | |||
FQ212377.1 | - | 3 | 161 | |||
FQ223744.1 | - | 3 | 161 | |||
X15512.1 | - | 2 | 196 |
Gene Symbol: LOC100911905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752245.2 | XP_003752293.2 | 3 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
