siRNA Details
Gene Symbol: Azgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012826.2 | NP_036958.2 | 4 | 712 | ||
XM_006249003.2 | XP_006249065.1 | 6 | 893 | |||
XM_006249004.2 | XP_006249066.1 | 6 | 877 | |||
GenBank mRNA | BC105822.1 | - | 4 | 712 | 7371444 | |
D21058.1 | - | 4 | 711 | |||
FQ210978.1 | - | 731 | ||||
FQ219509.1 | - | 4 | 703 | |||
X75309.1 | - | 4 | 693 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
