siRNA Details
Gene Symbol: SLC2A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135585.1 | NP_001129057.1 | 4 | 671 | ||
NM_003039.2 | NP_003030.1 | 4 | 671 | |||
XM_005263491.3 | XP_005263548.1 | 5 | 469 | |||
XM_017002133.1 | XP_016857622.1 | 6 | 697 | |||
XM_017002134.1 | XP_016857623.1 | 6 | 663 | |||
XM_017002135.1 | XP_016857624.1 | 6 | 757 | |||
XM_017002136.1 | XP_016857625.1 | 6 | 733 | |||
XM_017002137.1 | XP_016857626.1 | 7 | 769 | |||
XM_017002138.1 | XP_016857627.1 | 7 | 745 | |||
XM_017002139.1 | XP_016857628.1 | 5 | 679 | |||
XM_017002140.1 | XP_016857629.1 | 6 | 600 | |||
XM_017002141.1 | XP_016857630.1 | 5 | 502 | |||
XM_017002142.1 | XP_016857631.1 | 2 | 242 | |||
GenBank mRNA | AF451839.1 | - | 2 | 184 | ||
AF479408.1 | - | 2 | 184 | |||
AK289849.1 | - | 4 | 427 | |||
AK290398.1 | - | 4 | 461 | |||
AK294368.1 | - | 4 | 457 | |||
AK300091.1 | - | 2 | 128 | |||
AK300477.1 | - | 2 | 247 | |||
AK302047.1 | - | 4 | 312 | |||
AK302057.1 | - | 7 | 595 | |||
AK302137.1 | - | 6 | 547 | |||
BC001692.2 | - | 4 | 430 | 2966459 | ||
BC001820.2 | - | 4 | 430 | 2966459 | ||
BC035878.1 | - | 4 | 458 | |||
M55531.1 | - | 4 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
