siRNA Details
Gene Symbol: Bmp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012827.2 | NP_036959.2 | 1226 | |||
XM_006251746.2 | XP_006251808.1 | 1988 | ||||
XM_006251747.2 | XP_006251809.1 | 1538 | ||||
XM_006251748.2 | XP_006251810.1 | 4 | 1520 | |||
XM_006251749.2 | XP_006251811.1 | 1262 | ||||
XM_006251751.2 | XP_006251813.1 | 1239 | ||||
XM_008770501.1 | XP_008768723.1 | 1352 | ||||
GenBank mRNA | AY184241.1 | - | 2 | 1088 | ||
BC078901.1 | - | 1226 | 7121295 | |||
Z22607.1 | - | 4 | 1488 |
