siRNA Details
Gene Symbol: Abcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012833.2 | NP_036965.1 | 27 | 4020 | ||
GenBank mRNA | D86086.1 | - | 27 | 3842 | ||
L49379.1 | - | 27 | 3901 | |||
X96393.1 | - | 27 | 3856 |
