siRNA Details
Gene Symbol: Dcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012841.1 | NP_036973.1 | 15 | 2607 | ||
XM_008772087.1 | XP_008770309.1 | 14 | 2259 | |||
XM_008772088.1 | XP_008770310.1 | 14 | 2259 | |||
GenBank mRNA | U68725.1 | - | 15 | 2607 |
