siRNA Details
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012842.1 | NP_036974.1 | 1136 | |||
GenBank mRNA | BC062030.1 | - | 1121 | 5623849 | ||
JX149564.1 | - | 980 | ||||
JX437943.1 | - | 980 | ||||
U04842.1 | - | 1136 |
Gene Symbol: LOC100910178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760814.1 | XP_008759036.1 | 4, 5 | 1147 | ||
XM_008760815.1 | XP_008759037.1 | 4, 5 | 1146 | |||
XM_008760816.1 | XP_008759038.1 | 4, 5 | 907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
