siRNA Details
Gene Symbol: Fnta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012847.1 | NP_036979.1 | 8 | 934 | ||
GenBank mRNA | BC085758.1 | - | 8 | 884 | 7189836 | |
M81225.1 | - | 8 | 934 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
