siRNA Details
Gene Symbol: Fnta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012847.1 | NP_036979.1 | 8 | 926 | ||
GenBank mRNA | BC085758.1 | - | 8 | 876 | 7189836 | |
M81225.1 | - | 8 | 926 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fnta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012847.1 | NP_036979.1 | 8 | 926 | ||
GenBank mRNA | BC085758.1 | - | 8 | 876 | 7189836 | |
M81225.1 | - | 8 | 926 |