siRNA Details
Gene Symbol: Hsd17b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012851.2 | NP_036983.1 | 2 | 231 | ||
GenBank mRNA | BC086365.1 | - | 2 | 216 | 7305002 | |
X78811.1 | - | 2 | 192 | |||
X97754.1 | - | 2 | 232 |

More Information
Other Species Targeted

Gene Ontology Categories:
