siRNA Details
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012853.1 | NP_036985.1 | 4, 5 | 522 | ||
XM_006254793.2 | XP_006254855.1 | 5, 6 | 1061 | |||
XM_008772089.1 | XP_008770311.1 | 3, 4 | 2187 | |||
XM_008772090.1 | XP_008770312.1 | 3, 4 | 2187 | |||
GenBank mRNA | AJ011370.1 | - | 5, 6 | 602 | ||
AM712909.1 | - | 4, 5 | 502 | |||
AM712910.1 | - | 4, 5 | 502 | |||
AM712911.1 | - | 4, 5 | 502 | |||
U20906.1 | - | 4, 5 | 522 | |||
U20907.1 | - | 4, 5 | 522 | |||
Z48153.1 | - | 2, 3 | 220 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
