siRNA Details
Gene Symbol: Il10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012854.2 | NP_036986.2 | 3 | 272 | ||
XM_006249712.2 | XP_006249774.1 | 3 | 611 | |||
XM_008769426.1 | XP_008767648.1 | 3 | 611 | |||
GenBank mRNA | L02926.1 | - | 3 | 272 | ||
X60675.1 | - | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
