siRNA Details
Gene Symbol: Nfyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031553.2 | NP_113741.1 | 4 | 354 | ||
XM_006241196.2 | XP_006241258.1 | 4 | 372 | |||
GenBank mRNA | BC089791.1 | - | 4 | 354 | 7380395 | |
M55045.1 | - | 3 | 223 | |||
M60617.1 | - | 1 | 27 |

More Information
Other Species Targeted

Gene Ontology Categories:
