siRNA Details
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012866.2 | NP_036998.1 | 10 | 1146 | ||
XM_006238785.2 | XP_006238847.1 | 10 | 1119 | |||
GenBank mRNA | BC093619.1 | - | 10 | 1146 | 7314492 | |
U17607.1 | - | 10 | 1034 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
