siRNA Details
Gene Symbol: Npr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012868.1 | NP_037000.1 | 3 | 933 | ||
XM_006232045.2 | XP_006232107.1 | 3 | 1517 | |||
GenBank mRNA | L27339.1 | - | 3 | 933 | ||
X75605.1 | - | 3 | 954 | |||
X78595.1 | - | 779 |

More Information
Other Species Targeted

Gene Ontology Categories:
