siRNA Details
Gene Symbol: Tnfrsf11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012870.2 | NP_037002.1 | 3 | 656 | ||
GenBank mRNA | BC081830.1 | - | 3 | 656 | 7096099 | |
U94330.1 | - | 3 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
