siRNA Details
Gene Symbol: Rps29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012876.1 | NP_037008.1 | 3 | 256 | ||
GenBank mRNA | BC058150.1 | - | 3 | 254 | 6922365 | |
FQ209501.1 | - | 3 | 250 | |||
FQ210507.1 | - | 3 | 250 | |||
FQ211411.1 | - | 3 | 265 | |||
FQ211740.1 | - | 3 | 250 | |||
FQ216880.1 | - | 3 | 250 | |||
FQ217140.1 | - | 3 | 250 | |||
FQ217213.1 | - | 3 | 250 | |||
FQ217476.1 | - | 3 | 250 | |||
FQ218006.1 | - | 3 | 250 | |||
FQ218037.1 | - | 3 | 251 | |||
FQ218769.1 | - | 3 | 250 | |||
FQ221030.1 | - | 3 | 250 | |||
FQ221251.1 | - | 3 | 250 | |||
FQ221298.1 | - | 3 | 250 | |||
FQ221307.1 | - | 3 | 247 | |||
FQ221360.1 | - | 3 | 250 | |||
FQ221363.1 | - | 3 | 250 | |||
FQ221457.1 | - | 3 | 250 | |||
FQ221523.1 | - | 3 | 250 | |||
FQ221587.1 | - | 3 | 251 | |||
FQ221640.1 | - | 3 | 250 | |||
FQ221661.1 | - | 3 | 250 | |||
FQ221707.1 | - | 3 | 250 | |||
FQ221875.1 | - | 3 | 250 | |||
FQ221942.1 | - | 3 | 250 | |||
FQ222143.1 | - | 3 | 250 | |||
FQ222293.1 | - | 3 | 250 | |||
FQ222368.1 | - | 3 | 250 | |||
FQ222566.1 | - | 3 | 250 | |||
FQ222711.1 | - | 3 | 250 | |||
FQ222937.1 | - | 3 | 250 | |||
FQ223071.1 | - | 3 | 250 | |||
FQ223222.1 | - | 3 | 250 | |||
FQ223289.1 | - | 3 | 250 | |||
FQ223378.1 | - | 3 | 250 | |||
FQ223402.1 | - | 3 | 250 | |||
FQ223735.1 | - | 3 | 287 | |||
FQ223885.1 | - | 3 | 250 | |||
FQ223975.1 | - | 3 | 250 | |||
FQ224325.1 | - | 3 | 250 | |||
FQ224400.1 | - | 3 | 250 | |||
FQ224476.1 | - | 3 | 250 | |||
FQ224629.1 | - | 3 | 257 | |||
FQ224661.1 | - | 3 | 251 | |||
FQ224820.1 | - | 3 | 250 | |||
FQ228434.1 | - | 3 | 250 | |||
FQ228730.1 | - | 3 | 250 | |||
X59051.1 | - | 3 | 256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
