siRNA Details
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012881.2 | NP_037013.2 | 6 | 453 | ||
XM_008769996.1 | XP_008768218.1 | 6 | 526 | |||
GenBank mRNA | AB001382.1 | - | 5 | 347 | ||
BC078874.1 | - | 6 | 433 | 7110677 | ||
CV102664.1 | - | 6 | 453 | |||
FQ213473.1 | - | 6 | 442 | |||
FQ222192.1 | - | 6 | 469 | |||
M14656.1 | - | 6 | 426 | |||
M99252.1 | - | 6 | 462 |
