siRNA Details
Gene Symbol: Timp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012886.2 | NP_037018.2 | 4, 5 | 720 | ||
XM_006241162.2 | XP_006241224.1 | 4, 5 | 749 | |||
GenBank mRNA | BC097335.1 | - | 4, 5 | 720 | 7443686 | |
U27201.1 | - | 4, 5 | 435 |

More Information
Other Species Targeted

Gene Ontology Categories:
