siRNA Details
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012888.1 | NP_037020.1 | 7 | 642 | ||
XM_008764742.1 | XP_008762964.1 | 7 | 1214 | |||
GenBank mRNA | M34842.1 | - | 7 | 642 |

More Information
Other Species Targeted

Gene Ontology Categories:
