siRNA Details
Gene Symbol: Acadvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012891.2 | NP_037023.1 | 8, 9 | 764 | ||
GenBank mRNA | BC087036.1 | - | 8, 9 | 785 | 7193535 | |
D30647.1 | - | 8, 9 | 766 | |||
FQ210754.1 | - | 8, 9 | 813 | |||
FQ219329.1 | - | 8, 9 | 809 |
