siRNA Details
Gene Symbol: Arrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012911.1 | NP_037043.1 | 8 | 703 | ||
GenBank mRNA | BC087578.1 | - | 8 | 575 | 7311618 | |
M91590.1 | - | 8 | 703 |

More Information
Other Species Targeted

Gene Ontology Categories:
