siRNA Details
Gene Symbol: Arrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012911.1 | NP_037043.1 | 14 | 1315 | ||
GenBank mRNA | BC087578.1 | - | 14 | 1187 | 7311618 | |
M91590.1 | - | 14 | 1315 | |||
U03627.1 | - | 4 | 242 |
