siRNA Details
Gene Symbol: Atf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012912.2 | NP_037044.1 | 2 | 261 | ||
GenBank mRNA | BC078903.1 | - | 2 | 261 | 7100767 | |
M63282.1 | - | 2 | 266 |

More Information
Other Species Targeted

Gene Ontology Categories:
