siRNA Details
Gene Symbol: Atpif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012915.2 | NP_037047.2 | 3 | 315 | ||
GenBank mRNA | BC126065.1 | - | 3 | 280 | 7442425 | |
D13122.1 | - | 3 | 315 | |||
FQ221416.1 | - | 3 | 300 | |||
FQ229412.1 | - | 3 | 277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
