siRNA Details
Gene Symbol: Cacna1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012918.3 | NP_037050.2 | 32 | 4883 | ||
XM_008772347.1 | XP_008770569.1 | 33 | 5226 | |||
XM_008772348.1 | XP_008770570.1 | 33 | 5226 | |||
XM_008772349.1 | XP_008770571.1 | 33 | 5223 | |||
XM_008772350.1 | XP_008770572.1 | 32 | 5220 | |||
XM_008772351.1 | XP_008770573.1 | 32 | 5214 | |||
XM_008772352.1 | XP_008770574.1 | 33 | 5226 | |||
XM_008772353.1 | XP_008770575.1 | 33 | 5226 | |||
XM_008772354.1 | XP_008770576.1 | 33 | 5226 | |||
XM_008772355.1 | XP_008770577.1 | 32 | 5220 | |||
XM_008772356.1 | XP_008770578.1 | 32 | 5214 | |||
XM_008772357.1 | XP_008770579.1 | 31 | 5208 | |||
XM_008772358.1 | XP_008770580.1 | 31 | 5208 | |||
XM_008772359.1 | XP_008770581.1 | 33 | 5226 | |||
XM_008772360.1 | XP_008770582.1 | 33 | 5226 | |||
XM_008772361.1 | XP_008770583.1 | 35 | 5653 | |||
GenBank mRNA | AF051526.1 | - | 3 | 188 | ||
AF051527.1 | - | 3 | 188 | |||
AM040230.1 | - | 3 | 323 | |||
AM040231.1 | - | 6 | 578 | |||
AM040232.1 | - | 4 | 340 | |||
AM040233.1 | - | 4 | 285 | |||
AM040234.1 | - | 3 | 196 | |||
M64373.1 | - | 32 | 4883 | |||
M99222.1 | - | 8 | 831 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001252060, NM_001252061, NM_007578, NM_001252059, AY714490, and others.
Gene Ontology Categories:
