siRNA Details
Gene Symbol: Cacna2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110847.1 | NP_001104317.1 | 27 | 2245 | ||
NM_001110848.1 | NP_001104318.1 | 27 | 2248 | |||
NM_012919.3 | NP_037051.2 | 28 | 2554 | |||
XM_006235981.2 | XP_006236043.1 | 29 | 2601 | |||
XM_006235982.2 | XP_006236044.1 | 28 | 2580 | |||
XM_006235983.2 | XP_006236045.1 | 28 | 2529 | |||
GenBank mRNA | AF286488.1 | - | 28 | 2555 | ||
AF400662.1 | - | 27 | 2245 | |||
AF486276.1 | - | 27 | 2248 | |||
M86621.1 | - | 28 | 2423 |

More Information
Other Species Targeted

Gene Ontology Categories:
