siRNA Details
Gene Symbol: Casp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012922.2 | NP_037054.1 | 4, 5 | 364 | ||
XM_006253130.2 | XP_006253192.1 | 4, 5 | 408 | |||
GenBank mRNA | BC081854.1 | - | 4, 5 | 364 | 7104685 | |
U34685.1 | - | 2, 3 | 222 | |||
U49930.1 | - | 3, 4 | 334 | |||
U84410.1 | - | 3, 4 | 338 |

More Information
Other Species Targeted

Gene Ontology Categories:
