siRNA Details
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012924.2 | NP_037056.2 | 6 | 862 | ||
XM_008762059.1 | XP_008760281.1 | 6 | 1011 | |||
GenBank mRNA | AF014365.1 | - | 1 | 80 | ||
U96138.1 | - | 6 | 862 |
