siRNA Details
Gene Symbol: Cpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012930.1 | NP_037062.1 | 5 | 1774 | ||
GenBank mRNA | J05470.1 | - | 5 | 1774 | ||
U88295.1 | - | 5 | 1712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
